Assay Details
Target Gene Details
Entrez Gene ID: | 101929341 |
Gene Name: | uncharacterized LOC101929341 |
Gene Aliases: |
- |
Location: |
Chr.8:52194362-52199552 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929341 | NR_134310.1 | 1 | 122 |
Target Gene Details
Entrez Gene ID: | 9705 |
Gene Name: | ST18, C2H2C-type zinc finger |
Gene Aliases: |
NZF3, ZC2H2C3, ZC2HC10, ZNF387 |
Location: |
Chr.8:52110832-52409879 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
More Information
Set Membership: |
|

Genomic Map