Assay Details
Target Gene Details
Entrez Gene ID: | 2138 |
Gene Name: | EYA transcriptional coactivator and phosphatase 1 |
Gene Aliases: |
BOP, BOR, BOS1, OFC1 |
Location: |
Chr.8:71197433-71548130 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
More Information
Additional Information:
For this assay, SNP(s) [rs72622839] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map