Assay Details
Target Gene Details
Entrez Gene ID: | 6482 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 1 |
Gene Aliases: |
Gal-NAc6S, SIAT4A, SIATFL, ST3GalA, ST3GalA.1, ST3GalIA, ST3GalIA,1, ST3O |
Location: |
Chr.8:133454848-133572454 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL1 | NM_003033.3 | NP_003024.1 | ||
| NM_173344.2 | NP_775479.1 | |||
| XM_005251025.4 | XP_005251082.1 | |||
| XM_006716617.1 | XP_006716680.1 | |||
| XM_011517225.1 | XP_011515527.1 | |||
| XM_017013736.1 | XP_016869225.1 | |||
| XM_017013737.1 | XP_016869226.1 | |||
| AF059321.1 | AAC17874.1 | |||
| BC018357.1 | AAH18357.1 | |||
| BX648001.1 | ||||
| L13972.1 | AAC37574.1 | |||
| L29555.1 | AAA36612.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831464 | Chr.8:133270395 - 133462604 on Build GRCh38 | Loss |
|
| nsv6404 | Chr.8:133428623 - 133462273 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map