Assay Details
Target Gene Details
Entrez Gene ID: | 50807 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain 1 |
Gene Aliases: |
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P |
Location: |
Chr.8:130052104-130443848 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASAP1 | NM_001247996.1 | NP_001234925.1 | ||
| NM_018482.3 | NP_060952.2 | |||
| XM_005250925.1 | XP_005250982.1 | |||
| XM_006716563.3 | XP_006716626.1 | |||
| XM_006716564.1 | XP_006716627.1 | |||
| XM_006716565.3 | XP_006716628.1 | |||
| XM_006716566.1 | XP_006716629.1 | |||
| XM_011517052.2 | XP_011515354.1 | |||
| XM_011517053.1 | XP_011515355.1 | |||
| XM_017013467.1 | XP_016868956.1 | |||
| XM_017013468.1 | XP_016868957.1 | |||
| XM_017013469.1 | XP_016868958.1 | |||
| AB033075.1 | BAA86563.1 | |||
| BC137135.1 | ||||
| BX537768.1 | CAD97831.1 |
Target Gene Details
Entrez Gene ID: | 100507117 |
Gene Name: | ASAP1 intronic transcript 2 |
Gene Aliases: |
- |
Location: |
Chr.8:130082738-130084768 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASAP1-IT2 | NR_045385.1 | 1 | 1711 | |
| AK054840.1 | 1 | 1289 | BAB70813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465810 | Chr.8:130026863 - 130318766 on Build GRCh38 | Gain |
|
| nsv1018045 | Chr.8:129851398 - 130110398 on Build GRCh38 | Gain |
|
| esv3576608 | Chr.8:129860292 - 130116656 on Build GRCh38 | Gain |
|
| nsv612231 | Chr.8:129855966 - 130120346 on Build GRCh38 | Gain |
|
| nsv524349 | Chr.8:129856117 - 130120346 on Build GRCh38 | Gain |
|
| nsv612232 | Chr.8:129951013 - 130106412 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map