Assay Details
Target Gene Details
Entrez Gene ID: | 100302175 |
Gene Name: | microRNA 1207 |
Gene Aliases: |
MIRN1207, hsa-mir-1207 |
Location: |
Chr.8:128049152-128049238 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR1207 | NR_031612.1 |
Target Gene Details
Entrez Gene ID: | 5820 |
Gene Name: | Pvt1 oncogene (non-protein coding) |
Gene Aliases: |
LINC00079, MYC, NCRNA00079, onco-lncRNA-100 |
Location: |
Chr.8:127794533-128101253 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PVT1 | NR_003367.3 | |||
| D79458.1 |

Genomic Map