Assay Details
Target Gene Details
Entrez Gene ID: | 9134 |
Gene Name: | cyclin E2 |
Gene Aliases: |
CYCE2 |
Location: |
Chr.8:94880224-94896671 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNE2 | XM_017013958.1 | XP_016869447.1 |
Target Gene Details
Entrez Gene ID: | 137682 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 6 |
Gene Aliases: |
C8orf38 |
Location: |
Chr.8:94895271-95118496 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF6 | XM_011516835.2 | XP_011515137.1 | ||
| XM_017013030.1 | 1 | 461 | XP_016868519.1 | |
| XM_017013031.1 | 1 | 183 | XP_016868520.1 | |
| XM_017013032.1 | XP_016868521.1 | |||
| XM_017013035.1 | 1 | 132 | XP_016868524.1 | |
| XM_017013036.1 | 1 | 537 | XP_016868525.1 | |
| XM_017013039.1 | 1 | 132 | XP_016868528.1 |
More Information
Set Membership: |
|

Genomic Map