Assay Details
Target Gene Details
Entrez Gene ID: | 64798 |
Gene Name: | DEP domain containing MTOR-interacting protein |
Gene Aliases: |
DEP.6, DEPDC6 |
Location: |
Chr.8:119873655-120056202 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEPTOR | NM_001283012.1 | NP_001269941.1 | ||
| NM_022783.3 | NP_073620.2 | |||
| AK022490.1 | BAB14054.1 | |||
| AK023916.1 | BAB14723.1 | |||
| AK225168.1 | ||||
| AK225675.1 | ||||
| AK297822.1 | ||||
| AK315175.1 | ||||
| AL136678.1 | CAB66613.1 | |||
| BC012040.1 | AAH12040.1 | |||
| BC024746.1 | AAH24746.1 | |||
| DA622978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612111 | Chr.8:119624610 - 119982154 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75113975] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map