Assay Details
Target Gene Details
Entrez Gene ID: | 2915 |
Gene Name: | glutamate metabotropic receptor 5 |
Gene Aliases: |
GPRC1E, MGLUR5, PPP1R86, mGlu5 |
Location: |
Chr.11:88504576-89065971 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM5 | NM_000842.4 | NP_000833.1 | ||
| NM_001143831.2 | NP_001137303.1 | |||
| XM_006718828.3 | XP_006718891.1 | |||
| XM_011542792.1 | XP_011541094.1 | |||
| XM_017017627.1 | XP_016873116.1 | |||
| AK291362.1 | ||||
| AY608336.1 | AAT37960.1 | |||
| D28538.1 | BAA05891.1 | |||
| D28539.1 | BAA05892.1 | |||
| FJ588505.1 | ||||
| FJ588506.1 | ||||
| FJ588507.1 | ||||
| FJ668670.1 | ||||
| S64316.1 | AAD13954.1 |
Target Gene Details
Entrez Gene ID: | 100873989 |
Gene Name: | GRM5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:88504576-88524054 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM5-AS1 | NR_049724.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035387 | Chr.11:88442010 - 88772705 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map