Assay Details
Target Gene Details
Entrez Gene ID: | 51272 |
Gene Name: | Bet1 golgi vesicular membrane trafficking protein like |
Gene Aliases: |
BET1L1, GOLIM3, GS15, HSPC197 |
Location: |
Chr.11:202924-207422 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BET1L | NM_001098787.1 | NP_001092257.1 | ||
| NM_016526.4 | NP_057610.2 | |||
| AF234160.1 | AAF37877.1 | |||
| AK022958.1 | ||||
| BC008971.1 | AAH08971.1 | |||
| BC032779.2 | AAH32779.1 | |||
| DA875317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625061 | Chr.11:195766 - 233956 on Build GRCh38 | Gain |
|
| esv3625060 | Chr.11:194412 - 252166 on Build GRCh38 | Gain |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1012n100 | Chr.11:198510 - 427836 on Build GRCh38 | Gain |
|
| esv22231 | Chr.11:60059 - 216243 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs185553134] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map