Assay Details
Target Gene Details
Entrez Gene ID: | 6188 |
Gene Name: | ribosomal protein S3 |
Gene Aliases: |
S3 |
Location: |
Chr.11:75399491-75422302 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS3 | NM_001005.4 | NP_000996.2 | ||
| NM_001256802.1 | NP_001243731.1 | |||
| NM_001260506.1 | NP_001247435.1 | |||
| NM_001260507.1 | NP_001247436.1 | |||
| AA976344.1 | ||||
| AB062288.1 | BAB93471.1 | |||
| AK129794.1 | ||||
| AK130960.1 | ||||
| AK223048.1 | BAD96768.1 | |||
| BC003137.1 | AAH03137.1 | |||
| BC003577.1 | AAH03577.1 | |||
| BC013196.1 | AAH13196.1 | |||
| BC034149.1 | AAH34149.1 | |||
| BC071917.1 | AAH71917.1 | |||
| BC100284.1 | AAI00285.1 | |||
| BM928330.1 | ||||
| S42658.1 | AAB19349.2 | |||
| U14990.1 | AAB60336.1 | |||
| U14991.1 | AAB60337.1 | |||
| U14992.1 | AAB60338.1 | |||
| X55715.1 | CAA39248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825995 | Chr.11:75405036 - 75405500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map