Assay Details
Target Gene Details
Entrez Gene ID: | 11098 |
Gene Name: | protease, serine 23 |
Gene Aliases: |
SIG13, SPUVE, ZSIG13 |
Location: |
Chr.11:86791059-86952910 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS23 | NM_001293180.1 | NP_001280109.1 | ||
| DA396057.1 | ||||
| DA895063.1 | ||||
| DN917578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627099 | Chr.11:86660723 - 87045565 on Build GRCh38 | Gain |
|
| nsv1049157 | Chr.11:86698017 - 86793113 on Build GRCh38 | Gain |
|
| nsv428263 | Chr.11:86646948 - 86797438 on Build GRCh38 | Loss |
|
| nsv521505 | Chr.11:86727073 - 86804030 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map