Assay Details
Target Gene Details
Entrez Gene ID: | 28971 |
Gene Name: | adipogenesis associated Mth938 domain containing |
Gene Aliases: |
C11orf67, CK067, PTD015 |
Location: |
Chr.11:77821114-77901281 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAMDC | NM_001316957.1 | NP_001303886.1 | ||
| NM_001316958.1 | NP_001303887.1 | |||
| NR_133637.1 | ||||
| XM_017017620.1 | XP_016873109.1 | |||
| AF092136.1 | AAD40378.1 | |||
| BC016854.2 | AAH16854.1 | |||
| BP270361.1 | ||||
| BX646013.1 | ||||
| CN362204.1 |
Target Gene Details
Entrez Gene ID: | 92105 |
Gene Name: | integrator complex subunit 4 |
Gene Aliases: |
INT4, MST093 |
Location: |
Chr.11:77874491-77994849 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS4 | XM_017018560.1 | 23 | 3881 | XP_016874049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv524537 | Chr.11:77872220 - 77882175 on Build GRCh38 | Loss |
|
| nsv555453 | Chr.11:77866448 - 77907395 on Build GRCh38 | Loss |
|
| nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map