Assay Details
Target Gene Details
Entrez Gene ID: | 283237 |
Gene Name: | tetratricopeptide repeat domain 9C |
Gene Aliases: |
- |
Location: |
Chr.11:62728073-62738636 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC9C | NM_001318812.1 | NP_001305741.1 | ||
| NM_001318813.1 | NP_001305742.1 | |||
| NM_001318814.1 | NP_001305743.1 | |||
| NM_001318815.1 | NP_001305744.1 | |||
| NM_001318816.1 | NP_001305745.1 | |||
| NM_173810.3 | NP_776171.1 | |||
| AF451992.1 | AAP97691.1 | |||
| AF451993.1 | AAP97692.1 | |||
| BC032123.2 | AAH32123.1 | |||
| BC053665.1 | AAH53665.1 | |||
| BG720745.1 | ||||
| BQ050575.1 | ||||
| BX421356.2 | ||||
| CN430374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv348 | Chr.11:62706978 - 62752240 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map