Assay Details
Target Gene Details
Entrez Gene ID: | 26297 |
Gene Name: | secretion regulating guanine nucleotide exchange factor |
Gene Aliases: |
DELGEF, Gnefr |
Location: |
Chr.11:17788048-18013162 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERGEF | NM_012139.3 | NP_036271.1 | ||
| NR_104040.1 | ||||
| NR_104041.1 | ||||
| AF086056.1 | ||||
| AJ243950.1 | CAB60832.1 | |||
| AJ243951.1 | CAB60833.1 | |||
| AK292286.1 | ||||
| BC065375.1 | AAH65375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| nsv509390 | Chr.11:17726193 - 17855296 on Build GRCh38 | Insertion |
|
| nsv526501 | Chr.11:17794865 - 17802825 on Build GRCh38 | Loss |
|
| nsv832078 | Chr.11:17758956 - 17931974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map