Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC73 | NM_001008391.3 | NP_001008392.2 | ||
| XM_011520139.2 | XP_011518441.1 | |||
| AK128159.1 | BAC87304.1 | |||
| AK131470.1 | BAD18615.1 | |||
| BC062749.1 | AAH62749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159852 | Chr.11:32634097 - 32681188 on Build GRCh38 | Deletion |
|

Genomic Map