Assay Details
Target Gene Details
Entrez Gene ID: | 55031 |
Gene Name: | ubiquitin specific peptidase 47 |
Gene Aliases: |
TRFP |
Location: |
Chr.11:11841423-11959325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP47 | NM_001282659.1 | NP_001269588.1 | ||
| NM_017944.3 | NP_060414.3 | |||
| XM_005252997.1 | XP_005253054.1 | |||
| XM_017017953.1 | XP_016873442.1 | |||
| AK027362.1 | BAB55063.1 | |||
| AK092290.1 | ||||
| AK127264.1 | ||||
| AK309240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054113 | Chr.11:11828552 - 11953953 on Build GRCh38 | Loss |
|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| nsv7681 | Chr.11:11860698 - 11905567 on Build GRCh38 | Deletion |
|
| nsv832067 | Chr.11:11777158 - 11976774 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map