Assay Details
Target Gene Details
Entrez Gene ID: | 117245 |
Gene Name: | HRAS like suppressor family member 5 |
Gene Aliases: |
HRLP5, HRSL5, RLP1, iNAT |
Location: |
Chr.11:63461404-63491208 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS5 | NM_001146728.1 | NP_001140200.1 | ||
| NM_001146729.1 | NP_001140201.1 | |||
| NM_054108.3 | NP_473449.1 | |||
| XM_011544751.2 | XP_011543053.1 | |||
| XM_011544752.2 | XP_011543054.1 | |||
| XM_011544753.1 | XP_011543055.1 | |||
| XM_017017172.1 | XP_016872661.1 | |||
| AB298804.1 | ||||
| AJ416558.1 | CAC94942.1 | |||
| AK055392.1 | ||||
| AK302312.1 | ||||
| BC034222.1 | AAH34222.1 |
More Information
Set Membership: |
|

Genomic Map