Assay Details
Target Gene Details
Entrez Gene ID: | 5920 |
Gene Name: | retinoic acid receptor responder 3 |
Gene Aliases: |
HRASLS4, HRSL4, PLA1/2-3, RIG1, TIG3 |
Location: |
Chr.11:63536801-63546458 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RARRES3 | NM_004585.4 | NP_004576.2 | ||
AB030815.1 | BAB08109.1 | |||
AB453252.1 | ||||
AF060228.1 | AAC84000.1 | |||
AF092922.1 | AAF02294.1 | |||
AK293357.1 | ||||
AK312110.1 | ||||
BC009678.1 | AAH09678.1 | |||
DA973023.1 | ||||
R55646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468588 | Chr.11:63465891 - 63552767 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
