Assay Details
Target Gene Details
Entrez Gene ID: | 55327 |
Gene Name: | lin-7 homolog C, crumbs cell polarity complex component |
Gene Aliases: |
LIN-7-C, LIN-7C, MALS-3, MALS3, VELI3 |
Location: |
Chr.11:27494418-27506779 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIN7C | NM_018362.3 | NP_060832.1 | ||
| AK002077.1 | BAA92072.1 | |||
| AK223156.1 | BAD96876.1 | |||
| AL833601.1 | ||||
| BC053907.1 | AAH53907.1 | |||
| DA231006.1 |
More Information
Set Membership: |
|

Genomic Map