Assay Details
Target Gene Details
Entrez Gene ID: | 5031 |
Gene Name: | pyrimidinergic receptor P2Y6 |
Gene Aliases: |
P2Y6 |
Location: |
Chr.11:73264503-73298625 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY6 | NM_001277205.1 | NP_001264134.1 | ||
| NM_001277206.1 | NP_001264135.1 | |||
| NM_001277207.1 | NP_001264136.1 | |||
| NM_176796.2 | NP_789766.1 | |||
| NM_176797.2 | NP_789767.1 | |||
| NM_176798.2 | NP_789768.1 | |||
| XM_011545076.2 | XP_011543378.1 | |||
| XM_011545077.2 | XP_011543379.1 | |||
| XM_011545079.2 | XP_011543381.1 | |||
| AF007891.1 | AAB80712.1 | |||
| AF007892.1 | AAB80713.1 | |||
| AF007893.1 | 1 | 397 | ||
| AL520218.3 | ||||
| AL525099.3 | ||||
| AX775903.1 | CAE11651.1 | |||
| BQ718871.1 | ||||
| BX337204.2 | ||||
| BX381745.2 | ||||
| BX397649.2 | ||||
| DR006998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv387 | Chr.11:73253689 - 73285467 on Build GRCh38 | Insertion |
|
| nsv832208 | Chr.11:73165139 - 73379958 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map