Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map