Assay Details
Target Gene Details
Entrez Gene ID: | 101928008 |
Gene Name: | uncharacterized LOC101928008 |
Gene Aliases: |
- |
Location: |
Chr.11:9839142-9929263 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928008 | NR_120539.1 | |||
| U80769.1 |
Target Gene Details
Entrez Gene ID: | 81846 |
Gene Name: | SET binding factor 2 |
Gene Aliases: |
CMT4B2, DENND7B, MTMR13 |
Location: |
Chr.11:9778667-10294216 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBF2 | NM_030962.3 | NP_112224.1 | ||
| XM_005253154.4 | XP_005253211.1 | |||
| XM_005253155.4 | XP_005253212.1 | |||
| XM_011520394.2 | XP_011518696.1 | |||
| XM_011520395.2 | XP_011518697.1 | |||
| XM_011520396.2 | XP_011518698.1 | |||
| XM_017018372.1 | XP_016873861.1 | |||
| XM_017018373.1 | XP_016873862.1 | |||
| XM_017018374.1 | XP_016873863.1 | |||
| XM_017018375.1 | XP_016873864.1 | |||
| XM_017018376.1 | XP_016873865.1 | |||
| XM_017018377.1 | XP_016873866.1 | |||
| AB051553.1 | BAB21857.1 | |||
| AY234241.1 | AAO62733.1 | |||
| BC101466.1 | AAI01467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744095 | Chr.11:9623376 - 10133785 on Build GRCh38 | Deletion |
|
| nsv1053369 | Chr.11:9626290 - 9854507 on Build GRCh38 | Gain |
|
| esv3579359 | Chr.11:9805474 - 9871834 on Build GRCh38 | Loss |
|
| esv2762895 | Chr.11:9827964 - 9933002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map