Assay Details
Target Gene Details
Entrez Gene ID: | 6484 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 4 |
Gene Aliases: |
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ |
Location: |
Chr.11:126355645-126459601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL4 | NM_001254757.1 | NP_001241686.1 | ||
| NM_006278.2 | NP_006269.1 | |||
| XM_017018134.1 | XP_016873623.1 | |||
| AB009393.1 | ||||
| AL520639.3 | ||||
| AL529887.3 | ||||
| BI259855.1 | ||||
| DA210431.1 | ||||
| X74570.1 | CAA52662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv235e214 | Chr.11:126365411 - 126381998 on Build GRCh38 | Gain |
|
| esv24669 | Chr.11:126365316 - 126381012 on Build GRCh38 | Gain |
|
| esv2745215 | Chr.11:126375566 - 126396937 on Build GRCh38 | Deletion |
|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
| esv2762951 | Chr.11:126366419 - 126380525 on Build GRCh38 | Gain |
|
| esv3579953 | Chr.11:126370213 - 126387884 on Build GRCh38 | Loss |
|
| esv3892106 | Chr.11:126372802 - 126378316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map