Assay Details
Target Gene Details
Entrez Gene ID: | 627 |
Gene Name: | brain derived neurotrophic factor |
Gene Aliases: |
ANON2, BULN2 |
Location: |
Chr.11:27654893-27722058 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Gene Details
Entrez Gene ID: | 497258 |
Gene Name: | BDNF antisense RNA |
Gene Aliases: |
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049 |
Location: |
Chr.11:27506852-27698171 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BDNF-AS | NR_033312.1 | 9 | 1493 | |
| NR_033313.1 | 6 | 736 | ||
| NR_033315.1 | 7 | 894 | ||
| AJ011597.1 | 1 | 151 | ||
| AY054401.3 | 9 | 1494 | ||
| AY054402.3 | 6 | 736 | ||
| AY513486.3 | 7 | 894 | ||
| EF689022.1 | 5 | 519 | ||
| EF689024.1 | 5 | 802 | ||
| EF689026.1 | 5 | 726 | ||
| EF689028.1 | 5 | 644 | ||
| EF689030.1 | 7 | 968 | ||
| EF689032.1 | 7 | 892 | ||
| EF689034.1 | 7 | 810 | ||
| EF689036.1 | 6 | 899 | ||
| EF689038.1 | 6 | 823 | ||
| EF689040.1 | 6 | 741 | ||
| EF689042.1 | 8 | 968 |
More Information
Set Membership: |
|

Genomic Map