Assay Details
Target Gene Details
Entrez Gene ID: | 55107 |
Gene Name: | anoctamin 1 |
Gene Aliases: |
DOG1, ORAOV2, TAOS2, TMEM16A |
Location: |
Chr.11:70078302-70189545 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map