Assay Details
Target Gene Details
Entrez Gene ID: | 399972 |
Gene Name: | ST3GAL4 antisense RNA 1 (head to head) |
Gene Aliases: |
DCPS-AS1 |
Location: |
Chr.11:126341716-126355587 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL4-AS1 | NR_033839.1 | |||
| AK096370.1 | 3 | 1475 | ||
| BF343672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|

Genomic Map