Assay Details
Target Gene Details
Entrez Gene ID: | 7003 |
Gene Name: | TEA domain transcription factor 1 |
Gene Aliases: |
AA, NTEF-1, REF1, TCF-13, TCF13, TEAD-1, TEF-1 |
Location: |
Chr.11:12674422-12944737 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEAD1 | NM_021961.5 | NP_068780.2 | ||
| AI199443.1 | ||||
| BC115398.1 | ||||
| DR000961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825769 | Chr.11:12647740 - 12703894 on Build GRCh38 | Gain |
|
| esv3625395 | Chr.11:12409149 - 13018985 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116605334] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map