Assay Details
Target Gene Details
Entrez Gene ID: | 100009613 |
Gene Name: | ANO1 antisense RNA 2 (head to head) |
Gene Aliases: |
C11orf78, NCRNA00224 |
Location: |
Chr.11:70072434-70075348 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANO1-AS2 | NR_103835.1 | |||
| AW592641.1 | ||||
| EF419887.1 |

Genomic Map