Assay Details
Target Gene Details
Entrez Gene ID: | 25988 |
Gene Name: | histone H4 transcription factor |
Gene Aliases: |
HiNF-P, MIZF, ZNF743 |
Location: |
Chr.11:119121523-119135055 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HINFP | NM_001243259.1 | NP_001230188.1 | ||
| NM_015517.4 | NP_056332.2 | |||
| NM_198971.2 | NP_945322.1 | |||
| XM_011542745.2 | XP_011541047.1 | |||
| XM_011542746.1 | XP_011541048.1 | |||
| XM_017017500.1 | XP_016872989.1 | |||
| XM_017017501.1 | XP_016872990.1 | |||
| XM_017017502.1 | XP_016872991.1 | |||
| XM_017017504.1 | XP_016872993.1 | |||
| XM_017017506.1 | XP_016872995.1 | |||
| XM_017017507.1 | XP_016872996.1 | |||
| XM_017017508.1 | XP_016872997.1 | |||
| XM_017017509.1 | XP_016872998.1 | |||
| AK056362.1 | ||||
| AK300288.1 | ||||
| AK301452.1 | ||||
| BC012856.1 | AAH12856.1 | |||
| BC017234.2 | AAH17234.1 | |||
| BF726036.1 | ||||
| BG821695.1 | ||||
| DC326345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv514 | Chr.11:119114552 - 119143217 on Build GRCh38 | Deletion |
|
| nsv468879 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73562680] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map