Assay Details
Target Gene Details
Entrez Gene ID: | 7352 |
Gene Name: | uncoupling protein 3 |
Gene Aliases: |
SLC25A9 |
Location: |
Chr.11:74000281-74009237 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP3 | NM_003356.3 | NP_003347.1 | ||
| NM_022803.2 | 6 | 1765 | NP_073714.1 | |
| AF001787.1 | AAC51369.1 | |||
| AF011449.1 | AAC51767.1 | |||
| AK092685.1 | 6 | 1412 | ||
| BC008392.1 | AAH08392.1 | |||
| KU178371.1 | ||||
| KU178372.1 | ||||
| U84763.1 | AAC51367.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117552079] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map