Assay Details
Target Gene Details
Entrez Gene ID: | 51317 |
Gene Name: | PHD finger protein 21A |
Gene Aliases: |
BHC80, BM-006 |
Location: |
Chr.11:45929319-46121434 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF21A | NM_001101802.1 | 18 | 7146 | NP_001095272.1 |
| NM_016621.3 | 18 | 7008 | NP_057705.3 | |
| XM_011520158.2 | 19 | 7051 | XP_011518460.1 | |
| XM_017017893.1 | 20 | 7088 | XP_016873382.1 | |
| XM_017017901.1 | 18 | 6895 | XP_016873390.1 | |
| AL359608.1 | 1 | 1973 | ||
| BX648236.1 | 10 | 8790 | CAH10542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516442 | Chr.11:45878411 - 45961834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map