Assay Details
Target Gene Details
Entrez Gene ID: | 272 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.11:10450321-10507579 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMPD3 | NM_000480.2 | NP_000471.1 | ||
| NM_001025389.1 | NP_001020560.1 | |||
| NM_001025390.1 | NP_001020561.1 | |||
| NM_001172430.1 | NP_001165901.1 | |||
| NM_001172431.1 | NP_001165902.1 | |||
| AK289998.1 | ||||
| AK294444.1 | ||||
| AK295046.1 | ||||
| AK299113.1 | ||||
| AK301507.1 | ||||
| AK302970.1 | ||||
| AK308181.1 | ||||
| AK311436.1 | ||||
| AK311607.1 | ||||
| AK311711.1 | ||||
| AK313039.1 | ||||
| BC126118.1 | ||||
| D12775.1 | BAA02240.1 | |||
| DC334250.1 | ||||
| M84720.1 | AAA58365.1 | |||
| M84721.1 | AAA58366.1 | |||
| M84722.1 | AAA58367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625358 | Chr.11:10435119 - 10512909 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114400965] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map