Assay Details
Target Gene Details
Entrez Gene ID: | 23387 |
Gene Name: | SIK family kinase 3 |
Gene Aliases: |
L19, QSK, SIK-3 |
Location: |
Chr.11:116843402-117098453 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIK3 | NM_001281748.1 | NP_001268677.1 | ||
| NM_001281749.1 | NP_001268678.1 | |||
| NM_025164.4 | NP_079440.3 | |||
| XM_005271481.2 | XP_005271538.1 | |||
| XM_005271482.4 | XP_005271539.2 | |||
| XM_005271484.3 | XP_005271541.2 | |||
| XM_011542721.2 | XP_011541023.1 | |||
| XM_011542722.1 | XP_011541024.1 | |||
| XM_011542723.2 | XP_011541025.1 | |||
| XM_011542724.2 | XP_011541026.1 | |||
| XM_011542725.2 | XP_011541027.1 | |||
| XM_011542726.1 | XP_011541028.1 | |||
| XM_017017424.1 | XP_016872913.1 | |||
| XM_017017425.1 | XP_016872914.1 | |||
| XM_017017426.1 | XP_016872915.1 | |||
| XM_017017427.1 | XP_016872916.1 | |||
| AB209327.1 | BAD92564.1 | |||
| BC128510.1 | ||||
| BC128511.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73023041] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map