Assay Details
Target Gene Details
Entrez Gene ID: | 100130987 |
Gene Name: | uncharacterized LOC100130987 |
Gene Aliases: |
- |
Location: |
Chr.11:67317839-67391687 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130987 | NR_024469.1 | |||
| AK096155.1 | ||||
| DA400240.1 | ||||
| DB058546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468603 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|

Genomic Map