Assay Details
Target Gene Details
Entrez Gene ID: | 5140 |
Gene Name: | phosphodiesterase 3B |
Gene Aliases: |
HcGIP1, cGIPDE1 |
Location: |
Chr.11:14643691-14872059 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE3B | NM_000922.3 | NP_000913.2 | ||
| XM_006718249.3 | XP_006718312.1 | |||
| XM_017017911.1 | XP_016873400.1 | |||
| XM_017017912.1 | XP_016873401.1 | |||
| AB209326.1 | BAD92563.1 | |||
| AY459346.1 | AAR24292.1 | |||
| BC136565.1 | ||||
| BC136566.1 | ||||
| BC144248.1 | ||||
| BC150307.1 | ||||
| X95520.1 | CAA64774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048243 | Chr.11:14498179 - 14790901 on Build GRCh38 | Gain |
|
| esv3625426 | Chr.11:14690430 - 14692669 on Build GRCh38 | Loss |
|
| nsv1042405 | Chr.11:14668177 - 14714291 on Build GRCh38 | Loss |
|
| esv3578250 | Chr.11:14350967 - 14976647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map