Assay Details
Target Gene Details
Entrez Gene ID: | 51642 |
Gene Name: | mitochondrial ribosomal protein L48 |
Gene Aliases: |
CGI-118, HSPC290, L48MT, MRP-L48 |
Location: |
Chr.11:73787872-73864611 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL48 | NM_001318498.1 | NP_001305427.1 | ||
| NM_001318499.1 | NP_001305428.1 | |||
| NM_001318500.1 | NP_001305429.1 | |||
| NM_016055.5 | NP_057139.1 | |||
| NR_134658.1 | ||||
| NR_134659.1 | ||||
| NR_134660.1 | ||||
| NR_134661.1 | ||||
| XM_017017919.1 | XP_016873408.1 | |||
| AF151876.1 | AAD34113.1 | |||
| AF161408.1 | AAF28968.1 | |||
| AK297746.1 | ||||
| AK309219.1 | ||||
| AK310005.1 | ||||
| BC009764.1 | AAH09764.1 | |||
| BI549412.1 | ||||
| BM817807.1 | ||||
| BM828996.1 | ||||
| BP210564.1 | ||||
| BU600573.1 | ||||
| BU932890.1 | ||||
| BX354271.2 | ||||
| CF541319.1 | ||||
| DA549653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509430 | Chr.11:73794575 - 73860940 on Build GRCh38 | Insertion |
|
| nsv391 | Chr.11:73797814 - 73832998 on Build GRCh38 | Insertion |
|
| nsv1045149 | Chr.11:73603520 - 73870863 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map