Assay Details
Target Gene Details
Entrez Gene ID: | 83700 |
Gene Name: | junctional adhesion molecule 3 |
Gene Aliases: |
JAM-2, JAM-3, JAM-C, JAMC |
Location: |
Chr.11:134068925-134151757 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAM3 | NM_001205329.1 | NP_001192258.1 | ||
| NM_032801.4 | NP_116190.3 | |||
| AF356518.1 | AAK27221.1 | |||
| AF448478.1 | AAM20925.1 | |||
| AJ344431.1 | CAC69845.1 | |||
| AJ416101.1 | CAC94776.1 | |||
| AK074769.1 | BAC11195.1 | |||
| AK075309.1 | BAC11538.1 | |||
| AK125071.1 | ||||
| AK225666.1 | ||||
| AY358335.1 | AAQ88701.1 | |||
| BC010690.1 | AAH10690.1 | |||
| BC012147.1 | AAH12147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map