Assay Details
Target Gene Details
Entrez Gene ID: | 50862 |
Gene Name: | ring finger protein 141 |
Gene Aliases: |
ZFP26, ZNF230 |
Location: |
Chr.11:10511678-10541227 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF141 | NM_016422.3 | NP_057506.2 | ||
| AF214680.1 | AAF30180.1 | |||
| AK289764.1 | ||||
| AK309367.1 | ||||
| BC018104.1 | AAH18104.1 | |||
| BT006662.1 | AAP35308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044011 | Chr.11:10507806 - 10542903 on Build GRCh38 | Gain |
|
| nsv819964 | Chr.11:10515707 - 10523988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map