Assay Details
Target Gene Details
Entrez Gene ID: | 847 |
Gene Name: | catalase |
Gene Aliases: |
- |
Location: |
Chr.11:34438925-34472060 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAT | NM_001752.3 | NP_001743.1 | ||
| AK291585.1 | ||||
| AK301577.1 | ||||
| AK310184.1 | ||||
| AK315350.1 | ||||
| AY028632.1 | AAK29181.1 | |||
| BC110398.1 | ||||
| BC112217.1 | AAI12218.1 | |||
| BC112219.1 | AAI12220.1 | |||
| X04076.1 | CAA27717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832114 | Chr.11:34402881 - 34586304 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map