Assay Details
Target Gene Details
Entrez Gene ID: | 2132 |
Gene Name: | exostosin glycosyltransferase 2 |
Gene Aliases: |
SOTV, SSMS |
Location: |
Chr.11:44095549-44245430 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXT2 | NM_000401.3 | NP_000392.3 | ||
| NM_001178083.1 | NP_001171554.1 | |||
| NM_207122.1 | NP_997005.1 | |||
| XM_011519950.1 | XP_011518252.1 | |||
| XM_011519951.1 | XP_011518253.1 | |||
| AK296713.1 | ||||
| AK312375.1 | ||||
| BC010058.1 | AAH10058.1 | |||
| BC068545.1 | AAH68545.1 | |||
| BX648142.1 | ||||
| U62740.1 | AAB07008.1 | |||
| U64511.1 | AAC50764.1 | |||
| U72263.1 | AAB62718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832139 | Chr.11:44063681 - 44277647 on Build GRCh38 | Loss |
|
| nsv1043523 | Chr.11:44082672 - 44137619 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map