Assay Details
Target Gene Details
Entrez Gene ID: | 2011 |
Gene Name: | microtubule affinity regulating kinase 2 |
Gene Aliases: |
EMK-1, EMK1, PAR-1, Par-1b, Par1b |
Location: |
Chr.11:63838928-63911020 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARK2 | NM_001039469.2 | NP_001034558.2 | ||
| NM_001163296.1 | NP_001156768.1 | |||
| NM_001163297.1 | NP_001156769.1 | |||
| NM_004954.4 | NP_004945.4 | |||
| XM_017017310.1 | XP_016872799.1 | |||
| AB188493.1 | BAD37141.1 | |||
| AK225766.1 | ||||
| AK307858.1 | ||||
| AY562187.1 | AAT67601.1 | |||
| BC008771.2 | AAH08771.2 | |||
| BC084540.1 | AAH84540.1 | |||
| BM724555.1 | ||||
| BX383836.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555180 | Chr.11:63748280 - 63945767 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114504508] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map