Assay Details
Target Gene Details
Entrez Gene ID: | 10480 |
Gene Name: | eukaryotic translation initiation factor 3 subunit M |
Gene Aliases: |
B5, GA17, PCID1, TANGO7, hfl-B5 |
Location: |
Chr.11:32583767-32602873 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3M | NM_001307929.1 | NP_001294858.1 | ||
| NM_006360.5 | NP_006351.2 | |||
| XM_011519840.2 | XP_011518142.1 | |||
| AF064603.1 | AAC17108.1 | |||
| AF277183.1 | AAK07542.1 | |||
| AK001768.1 | ||||
| AK131064.1 | ||||
| AK222564.1 | BAD96284.1 | |||
| AK292139.1 | ||||
| AK304378.1 | ||||
| AK307121.1 | ||||
| AK312512.1 | ||||
| AV713889.1 | ||||
| AY769947.1 | AAX12524.1 | |||
| BC019103.2 | AAH19103.1 | |||
| BC051292.1 | AAH51292.1 | |||
| CR450300.1 | CAG29296.1 |
More Information
Set Membership: |
|

Genomic Map