Assay Details
Target Gene Details
Entrez Gene ID: | 51773 |
Gene Name: | remodeling and spacing factor 1 |
Gene Aliases: |
HBXAP, RSF-1, XAP8, p325 |
Location: |
Chr.11:77660009-77872232 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSF1 | NM_016578.3 | NP_057662.3 | ||
| XM_005274051.2 | XP_005274108.1 | |||
| XM_017017923.1 | XP_016873412.1 | |||
| AF227948.2 | AAF61709.2 | |||
| AF380176.1 | AAK57515.1 | |||
| AK001268.1 | BAA91591.1 | |||
| BC046124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1234n100 | Chr.11:77572991 - 77724786 on Build GRCh38 | Gain |
|
| nsv555452 | Chr.11:77590296 - 77709269 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map