Assay Details
Target Gene Details
Entrez Gene ID: | 775 |
Gene Name: | calcium voltage-gated channel subunit alpha1 C |
Gene Aliases: |
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS |
Location: |
Chr.12:1969677-2697949 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Gene Details
Entrez Gene ID: | 100874369 |
Gene Name: | CACNA1C intronic transcript 2 |
Gene Aliases: |
- |
Location: |
Chr.12:2048352-2049463 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1C-IT2 | NR_046768.1 | 1 | 155 | |
| AI810102.1 | 2 | 435 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521838 | Chr.12:1951414 - 2190514 on Build GRCh38 | Gain |
|
| nsv826182 | Chr.12:2027229 - 2098908 on Build GRCh38 | Gain |
|
| nsv1040370 | Chr.12:1912229 - 2561851 on Build GRCh38 | Gain |
|
| nsv826183 | Chr.12:2038303 - 2148652 on Build GRCh38 | Gain |
|
| esv3580294 | Chr.12:2020191 - 2071938 on Build GRCh38 | Gain |
|
| esv3580037 | Chr.12:2047062 - 2054316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map