Assay Details
Target Gene Details
Entrez Gene ID: | 775 |
Gene Name: | calcium voltage-gated channel subunit alpha1 C |
Gene Aliases: |
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS |
Location: |
Chr.12:1969677-2697949 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Gene Details
Entrez Gene ID: | 100874370 |
Gene Name: | CACNA1C intronic transcript 3 |
Gene Aliases: |
- |
Location: |
Chr.12:2269776-2288745 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1C-IT3 | NR_046769.1 | |||
| AA421533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040370 | Chr.12:1912229 - 2561851 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map