Assay Details
Target Gene Details
Entrez Gene ID: | 29915 |
Gene Name: | host cell factor C2 |
Gene Aliases: |
HCF-2, HCF2 |
Location: |
Chr.12:104064458-104108954 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCFC2 | NM_013320.2 | NP_037452.1 | ||
| XM_017019241.1 | XP_016874730.1 | |||
| XM_017019242.1 | XP_016874731.1 | |||
| AF117210.1 | AAD27814.1 | |||
| AK223342.1 | BAD97062.1 | |||
| AK313466.1 | ||||
| BC033799.1 | AAH33799.1 | |||
| DB030069.1 |
More Information
Set Membership: |
|

Genomic Map