Assay Details
Target Gene Details
Entrez Gene ID: | 254013 |
Gene Name: | electron transfer flavoprotein beta subunit lysine methyltransferase |
Gene Aliases: |
C12orf72, ETFB-KMT, METTL20 |
Location: |
Chr.12:31647160-31669082 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFBKMT | NM_001135864.1 | NP_001129336.1 | ||
| AK290248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760267 | Chr.12:31473798 - 31756974 on Build GRCh38 | Gain+Loss |
|
| esv3629050 | Chr.12:31639465 - 31647416 on Build GRCh38 | Gain |
|
| esv3629051 | Chr.12:31646569 - 31648924 on Build GRCh38 | Loss |
|
| esv3581338 | Chr.12:31636660 - 31655272 on Build GRCh38 | Gain |
|
| nsv832365 | Chr.12:31489877 - 31655598 on Build GRCh38 | Loss |
|
| nsv662 | Chr.12:31642332 - 31676288 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map