Assay Details
Target Gene Details
Entrez Gene ID: | 27345 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 4 |
Gene Aliases: |
- |
Location: |
Chr.12:70366220-70434292 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB4 | NM_014505.5 | NP_055320.4 | ||
| XM_011538188.2 | XP_011536490.1 | |||
| AF160967.1 | AAF69805.1 | |||
| AF207992.1 | AAF28333.1 | |||
| AF215891.1 | AAF75596.1 | |||
| BC042446.2 | AAH42446.2 | |||
| BC050621.1 | AAH50621.2 | |||
| BU556960.1 |
More Information
Set Membership: |
|

Genomic Map