Assay Details
Target Gene Details
Entrez Gene ID: | 102724020 |
Gene Name: | uncharacterized LOC102724020 |
Gene Aliases: |
- |
Location: |
Chr.12:10015352-10030606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724020 | NR_120484.1 | |||
| BU570676.1 |

Genomic Map