Assay Details
Target Gene Details
| Entrez Gene ID: | 387893 | 
| Gene Name: | lysine methyltransferase 5A | 
| Gene Aliases: | PR-Set7, SET07, SET8, SETD8 | 
| Location: | Chr.12:123383778-123409357 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KMT5A | NM_001324504.1 | NP_001311433.1 | ||
| NM_001324505.1 | NP_001311434.1 | |||
| NM_020382.4 | NP_065115.3 | |||
| NR_136910.1 | ||||
| XM_006719394.3 | XP_006719457.1 | |||
| XM_011538333.2 | XP_011536635.1 | |||
| AK292645.1 | ||||
| AY102937.1 | AAM47033.1 | |||
| BC050346.1 | AAH50346.1 | |||
| HQ259683.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2746505 | Chr.12:123389563 - 123395053 on Build GRCh38 | Deletion |  KMT5A | 
| dgv44e19 | Chr.12:123389476 - 123395147 on Build GRCh38 | Loss |  KMT5A | 
| nsv826534 | Chr.12:123375063 - 123391807 on Build GRCh38 | Gain |  KMT5A | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	For this assay, SNP(s) [rs76801833] are located under a primer or probe  sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top